Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054066.4 | NP_473407.2 | 3, 4 | 356 | ||
GenBank mRNA | AF435950.1 | - | 3, 4 | 361 |