Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 2 | 294 | ||
NM_078478.5 | NP_510963.1 | 2 | 247 | |||
GenBank mRNA | AF412297.1 | - | 2 | 191 | ||
AK005636.1 | - | 2 | 191 | |||
AK093887.1 | - | 2 | 223 | |||
AK134592.1 | - | 2 | 177 | |||
AK146433.1 | - | 2 | 154 | |||
AK146853.1 | - | 2 | 210 | |||
AK151488.1 | - | 2 | 224 | |||
AK158904.1 | - | 2 | 237 | |||
AK159695.1 | - | 2 | 221 | |||
AK159724.1 | - | 2 | 221 | |||
AK164122.1 | - | 2 | 192 | |||
AK167970.1 | - | 2 | 218 | |||
AK168022.1 | - | 2 | 224 | |||
AK168678.1 | - | 2 | 295 | |||
AK168800.1 | - | 2 | 203 | |||
BC008622.1 | - | 2 | 225 | 3588758 | ||
BC010224.1 | - | 2 | 227 | 3500685 | ||
BY141813.1 | - | 2 | 253 |