Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 6 | 377 | ||
NM_207706.1 | NP_997589.1 | 6 | 377 | |||
GenBank mRNA | AF398884.1 | - | 5 | 282 | ||
AK033918.1 | - | 6 | 571 | |||
AK038097.1 | - | 6 | 368 | |||
AK038455.1 | - | 6 | 483 | |||
AK045428.1 | - | 6 | 378 | |||
AK047040.1 | - | 6 | 339 | |||
AK050823.1 | - | 6 | 340 | |||
AK053574.1 | - | 6 | 341 | |||
AK186343.1 | - | 2 | 100 | |||
BC023954.1 | - | 6 | 362 | 5320429 | ||
BC033418.1 | - | 5 | 307 |