Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080454.4 | NP_536702.3 | 2 | 1450 | ||
NM_175452.4 | NP_780661.2 | 2 | 1468 | |||
GenBank mRNA | AK046421.1 | - | 2 | 1469 | ||
AK046609.1 | - | 2 | 1451 | |||
BC113759.1 | - | 2 | 954 | 40046961 |