Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080458.2 | NP_536706.2 | 1 | 306 | ||
GenBank mRNA | AF391088.1 | - | 1 | 303 | ||
AK052587.1 | - | 1 | 307 | |||
AY373833.1 | - | 1 | 183 | |||
BC038287.1 | - | 1 | 357 | 4221532 |