Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8608
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
4
585
NM_001081752.2
NP_001075221.1
3
555
NM_001081753.2
NP_001075222.1
4
585
NM_001081754.2
NP_001075223.1
4
585
NM_001081755.2
NP_001075224.1
4
585
NM_001278912.1
NP_001265841.1
4
585
NM_001278913.1
NP_001265842.1
4
585
NM_001278914.1
NP_001265843.1
3
555
NM_001278915.1
NP_001265844.1
4
585
NM_001278916.1
NP_001265845.1
4
585
NM_001278917.1
NP_001265846.1
3
555
NM_001278918.1
NP_001265847.1
3
555
NM_001278939.1
NP_001265868.1
4
585
XM_005250187.1
XP_005250244.1
4
262
XM_005250188.1
XP_005250245.1
4
260
XM_011515868.1
XP_011514170.1
4
271
XM_011515869.1
XP_011514171.1
3
200
XM_011515870.1
XP_011514172.1
4
602
XM_011515871.1
XP_011514173.1
4
271
XM_011515872.1
XP_011514174.1
4
271
XM_011515873.1
XP_011514175.1
4
271
XM_011515874.1
XP_011514176.1
3
572
XM_011515875.1
XP_011514177.1
3
243
XM_011515876.1
XP_011514178.1
4
271
XM_011515877.1
XP_011514179.1
4
271
XM_017011813.1
XP_016867302.1
3
171
XM_017011814.1
XP_016867303.1
4
271
GenBank mRNA
AB208942.1
-
3
181
AK075494.1
-
4
262
AK075554.1
-
4
268
AK092232.1
-
3
170
AK094138.1
-
4
201
AK095990.1
-
4
205
AK122731.1
-
4
199
AK125511.1
-
3
215
AK130894.1
-
4
247
AK225659.1
-
4
199
AK304845.1
-
4
585
BC035570.1
-
4
190
BC050379.1
-
4
203
BC065566.1
-
4
216
6153564
BP284909.1
-
4
277
BX537939.1
-
4
205
BX538199.1
-
3
187
M36860.1
-
4
235
X52896.1
-
4
234
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline