Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054089.4 | NP_473430.3 | 3 | 344 | ||
GenBank mRNA | AF389908.1 | - | 3 | 213 | ||
AK030953.1 | - | 3 | 336 | |||
AK044895.1 | - | 3 | 345 | |||
AK050044.1 | - | 3 | 274 | |||
AK170949.1 | - | 2 | 1817 | |||
BC058183.1 | - | 3 | 348 | |||
BC075728.1 | - | 3 | 325 | 6513964 |