Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 12 | 1473 | ||
GenBank mRNA | AF244362.3 | - | 11 | 1108 | ||
AK088518.1 | - | 11 | 1169 | |||
AK216883.1 | - | 2 | 129 | |||
BC023956.1 | - | 12 | 1242 | 5321098 | ||
BC034059.1 | - | 6 | 477 | 4163877 | ||
BC096743.1 | - | 12 | 1496 | 30549276 |