Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177845.1 | NP_001171316.1 | 6 | 631 | ||
NM_001177846.1 | NP_001171317.1 | 6 | 613 | |||
NM_001177847.1 | NP_001171318.1 | 6 | 559 | |||
NM_054096.2 | NP_473437.2 | 6 | 623 | |||
GenBank mRNA | AF378130.1 | - | 6 | 532 | ||
AF410784.1 | - | 5 | 455 | |||
AK036431.1 | - | 6 | 552 | |||
AK080135.1 | - | 6 | 632 | |||
AK151117.1 | - | 6 | 561 | |||
AK168494.1 | - | 6 | 551 | |||
BC005584.1 | - | 3 | 289 | 3157913 | ||
BY743490.1 | - | 6 | 559 |