Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267625.1 | NP_001254554.1 | 4 | 411 | ||
NM_001267626.1 | NP_001254555.1 | 4 | 411 | |||
NM_058212.2 | NP_478119.1 | 4 | 408 | |||
GenBank mRNA | AF362750.1 | - | 4 | 380 | ||
AK012970.1 | - | 4 | 416 | |||
AK039011.1 | - | 4 | 392 | |||
BC048572.1 | - | 4 | 291 | 6705809 | ||
FJ151218.1 | - | 4 | 383 |