Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 2, 3 | 353 | ||
NM_078478.5 | NP_510963.1 | 2, 3 | 306 | |||
GenBank mRNA | AF412297.1 | - | 2, 3 | 250 | ||
AK005636.1 | - | 2, 3 | 250 | |||
AK093887.1 | - | 2, 3 | 282 | |||
AK134592.1 | - | 2, 3 | 236 | |||
AK146433.1 | - | 2, 3 | 213 | |||
AK146853.1 | - | 2, 3 | 269 | |||
AK151488.1 | - | 2, 3 | 283 | |||
AK158904.1 | - | 2, 3 | 296 | |||
AK159695.1 | - | 2, 3 | 280 | |||
AK159724.1 | - | 2, 3 | 280 | |||
AK164122.1 | - | 2, 3 | 251 | |||
AK167970.1 | - | 2, 3 | 277 | |||
AK168022.1 | - | 2, 3 | 283 | |||
AK168678.1 | - | 2, 3 | 354 | |||
AK168800.1 | - | 2, 3 | 262 | |||
BC008622.1 | - | 2, 3 | 284 | 3588758 | ||
BC010224.1 | - | 2, 3 | 286 | 3500685 | ||
BY141813.1 | - | 2, 3 | 312 |