Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 6 | 420 | ||
NM_207706.1 | NP_997589.1 | 6 | 420 | |||
GenBank mRNA | AF398884.1 | - | 5 | 325 | ||
AK032033.1 | - | 6 | 393 | |||
AK033918.1 | - | 6 | 614 | |||
AK038097.1 | - | 6 | 411 | |||
AK038455.1 | - | 6 | 526 | |||
AK045428.1 | - | 6 | 421 | |||
AK047040.1 | - | 6 | 382 | |||
AK050823.1 | - | 6 | 383 | |||
AK053574.1 | - | 6 | 384 | |||
AK129023.1 | - | 6 | 420 | |||
AK186343.1 | - | 2 | 143 | |||
BC023954.1 | - | 6 | 405 | 5320429 | ||
BC033418.1 | - | 5 | 350 |