Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080446.2 | NP_536694.2 | 2 | 401 | ||
GenBank mRNA | AB048542.1 | - | 2 | 326 | ||
AK043127.1 | - | 2 | 390 | |||
AK131122.1 | - | 2 | 423 | |||
BC068140.1 | - | 2 | 329 | 6843107 |