Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053255.3 | NP_444485.2 | 3 | 357 | ||
GenBank mRNA | AF308697.1 | - | 3 | 356 | ||
BC019371.1 | - | 3 | 327 | |||
BY286576.1 | - | 3 | 358 | |||
CV556353.1 | - | 1 | 92 |