Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 7, 8 | 1188 | ||
NM_001039087.1 | NP_001034176.1 | 8, 9 | 1302 | |||
NM_054050.2 | NP_473391.1 | 8, 9 | 1302 | |||
GenBank mRNA | AF348669.1 | - | 8, 9 | 1258 | ||
AK147468.1 | - | 8, 9 | 1303 | |||
AK147470.1 | - | 8, 9 | 1303 | |||
AK147506.1 | - | 8, 9 | 1271 | |||
AK147696.1 | - | 7, 8 | 1168 | |||
BC129881.1 | - | 7, 8 | 998 | 40110588 |