Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054053.4 | NP_473394.3 | 33 | 8012 | ||
GenBank mRNA | AF405692.1 | - | 29 | 8157 | ||
AF405693.1 | - | 23 | 5811 | |||
AF405694.1 | - | 8 | 2756 | |||
AF435926.1 | - | 33 | 8013 | |||
JQ013292.1 | - | 10 | 2673 |