Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 3 | 535 | ||
GenBank mRNA | AF117710.1 | - | 3 | 515 | ||
AF181832.1 | - | 3 | 428 | |||
AF181989.1 | - | 3 | 518 | |||
AF349114.1 | - | 3 | 520 | |||
AY136510.1 | - | 3 | 485 | |||
AY509193.1 | - | 3 | 535 | |||
BC007075.1 | - | 3 | 536 | 4292125 | ||
EU694432.1 | - | 3 | 532 | |||
M25113.1 | - | 3 | 485 | |||
V00497.1 | - | 3 | 535 | |||
V00500.1 | - | 3 | 485 |