Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054089.4 | NP_473430.3 | 5 | 1314 | ||
GenBank mRNA | AF389908.1 | - | 5 | 1183 | ||
AK030953.1 | - | 5 | 1304 | |||
AK044895.1 | - | 5 | 1315 | |||
BC058183.1 | - | 5 | 1318 | |||
BC075728.1 | - | 5 | 1295 | 6513964 |