Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 20 | 2691 | ||
GenBank mRNA | AF244362.3 | - | 19 | 2326 | ||
AK088518.1 | - | 19 | 2387 | |||
BC023956.1 | - | 20 | 2460 | 5321098 | ||
BC026415.1 | - | 2 | 199 | |||
BC034059.1 | - | 14 | 1695 | 4163877 | ||
BC096743.1 | - | 20 | 2714 | 30549276 |