Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080420.2 | NP_536345.2 | 9 | 1618 | ||
GenBank mRNA | AK018070.1 | - | 1 | 102 | ||
AK162992.1 | - | 1 | 66 | |||
BC016212.1 | - | 9 | 1625 | 4924899 |