Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000526.4 | NP_000517.2 | 1, 2 | 587 | ||
GenBank mRNA | BC002690.2 | - | 1, 2 | 572 | 3610489 | |
BC019097.2 | - | 1, 2 | 578 | 5013623 | ||
BC042437.1 | - | 1, 2 | 542 | 4776763 | ||
BC094830.1 | - | 1, 2 | 578 | 5443153 | ||
BT007186.1 | - | 1, 2 | 526 |