Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166031.2 | NP_001159503.2 | 4 | 1579 | ||
NM_080452.3 | NP_536700.3 | 4 | 1636 | |||
GenBank mRNA | AK051184.1 | - | 4 | 1636 | ||
AK156412.1 | - | 3 | 2079 | |||
AK160129.1 | - | 4 | 1629 | |||
AK165956.1 | - | 4 | 1643 | |||
AK172120.1 | - | 3 | 2099 | |||
AK172432.1 | - | 3 | 2079 | |||
BC033460.1 | - | 3 | 2085 | |||
BC048377.1 | - | 4 | 1998 | |||
BC060503.1 | - | 4 | 1612 | 6306321 |