Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080456.1 | NP_536704.1 | 3 | 408 | ||
GenBank mRNA | AB049943.1 | - | 3 | 423 | ||
AK165590.1 | - | 3 | 425 | |||
AK166374.1 | - | 3 | 423 | |||
AK167304.1 | - | 3 | 423 | |||
AK167316.1 | - | 3 | 423 | |||
AY061856.1 | - | 3 | 423 | |||
BC027271.1 | - | 3 | 408 | 3156971 |