Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080633.2 | NP_542364.1 | 2 | 151 | ||
GenBank mRNA | AK143917.1 | - | 2 | 130 | ||
AK144207.1 | - | 2 | 114 | |||
AK145511.1 | - | 2 | 131 | |||
AK150027.1 | - | 2 | 131 | |||
AK150954.1 | - | 3 | 115 | |||
AK165411.1 | - | 2 | 115 | |||
AK188132.1 | - | 1 | 50 | |||
AK189803.1 | - | 1 | 50 | |||
AK190452.1 | - | 1 | 50 | |||
AK192654.1 | - | 1 | 50 | |||
AK195361.1 | - | 1 | 50 | |||
AK196433.1 | - | 1 | 50 | |||
AK198495.1 | - | 1 | 50 | |||
AK211347.1 | - | 1 | 50 | |||
AK218063.1 | - | 1 | 50 | |||
BC004645.1 | - | 2 | 177 | 3158239 | ||
BC094462.1 | - | 2 | 110 | 4238599 | ||
BI987806.1 | - | 2 | 152 | |||
BY732916.1 | - | 2 | 134 |