Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080637.3 | NP_542368.2 | 2 | 203 | ||
GenBank mRNA | AF343565.1 | - | 1 | 92 | ||
AK006111.1 | - | 1 | 187 | |||
AK131722.1 | - | 2 | 204 | |||
AK138562.1 | - | 185 | ||||
BC049625.1 | - | 2 | 197 | 6704971 |