Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080793.5 | NP_542983.3 | 2 | 385 | ||
GenBank mRNA | AF448509.1 | - | 2 | 167 | ||
AK037895.1 | - | 1 | 1035 | |||
AK048924.1 | - | 2 | 222 | |||
AK129428.1 | - | 2 | 174 | |||
AK147316.1 | - | 2 | 441 | |||
AK147413.1 | - | 2 | 211 | |||
AK147422.1 | - | 2 | 386 | |||
AK147667.1 | - | 2 | 420 | |||
AK170161.1 | - | 2 | 229 | |||
BC050190.1 | - | 2 | 209 | 30007831 |