Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mydgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080837.2 | NP_543027.1 | 2, 3 | 227 | ||
GenBank mRNA | AK004074.1 | - | 2, 3 | 227 | ||
AK008289.1 | - | 2, 3 | 213 | |||
AK008854.1 | - | 2, 3 | 208 | |||
AK018016.1 | - | 2, 3 | 210 | |||
AK133947.1 | - | 211 | ||||
AK148083.1 | - | 2, 3 | 227 | |||
AK162834.1 | - | 2, 3 | 215 | |||
AY038184.1 | - | 2, 3 | 223 | |||
BC132127.1 | - | 2, 3 | 248 | 40130404 | ||
BC132129.1 | - | 2, 3 | 248 | 40130406 |