Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080852.2 | NP_543128.1 | 1 | 188 | ||
GenBank mRNA | AB055005.1 | - | 1 | 193 | ||
AJ417663.1 | - | 1 | 150 | |||
AK085644.1 | - | 1 | 177 |