Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080854.3 | NP_543130.2 | 2, 3 | 187 | ||
GenBank mRNA | AB054999.1 | - | 2, 3 | 167 | ||
BC125326.1 | - | 1, 2 | 120 | 40129841 | ||
BC131997.1 | - | 1, 2 | 120 | 40130274 |