Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130450.2 | NP_569717.1 | 2, 3 | 231 | ||
GenBank mRNA | AB072039.1 | - | 2, 3 | 178 | ||
AF480860.1 | - | 2, 3 | 150 | |||
AK029029.1 | - | 2, 3 | 233 | |||
AK087440.1 | - | 2, 3 | 251 | |||
AK165847.1 | - | 1, 2 | 255 | |||
AK167373.1 | - | 2, 3 | 197 | |||
AY053453.1 | - | 2, 3 | 212 | |||
BC040097.1 | - | 2, 3 | 173 | |||
BC051041.1 | - | 2, 3 | 228 | 4012676 | ||
BC098492.1 | - | 1, 2 | 228 | 30091954 | ||
BC100576.1 | - | 2, 3 | 173 | 30097278 |