Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130451.3 | NP_569718.1 | 2 | 762 | ||
GenBank mRNA | AK052156.1 | - | 2 | 729 | ||
AY029579.1 | - | 2 | 685 | |||
BC137917.1 | - | 2 | 680 | 8860936 | ||
BC137920.1 | - | 2 | 680 | 8860939 |