Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305102.1 | NP_001292031.1 | 3 | 503 | ||
NM_007271.3 | NP_009202.1 | 3 | 506 | |||
XM_006714988.3 | XP_006715051.1 | 3 | 886 | |||
XM_006714989.3 | XP_006715052.1 | 3 | 883 | |||
XM_017010226.1 | XP_016865715.1 | 3 | 466 | |||
GenBank mRNA | AF034188.1 | - | 2 | 169 | ||
AK312559.1 | - | 3 | 414 | |||
BC012085.1 | - | 3 | 460 | 4554884 | ||
BC095413.1 | - | 3 | 466 | 30337276 | ||
DA459946.1 | - | 3 | 503 | |||
Z35102.1 | - | 2 | 769 |