Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 2 | 268 | ||
GenBank mRNA | AF327431.1 | - | 2 | 140 | ||
AK089276.1 | - | 2 | 107 | |||
AK143217.1 | - | 2 | 269 | |||
AK157340.1 | - | 2 | 240 | |||
BC003901.1 | - | 2 | 213 | 3601310 |