Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205343.1 | NP_001192272.1 | 7 | 659 | ||
NM_130887.3 | NP_570957.2 | 7 | 659 | |||
GenBank mRNA | AF314171.1 | - | 7 | 632 | ||
AK053198.1 | - | 7 | 660 | |||
AK086767.1 | - | 7 | 640 | |||
AK138980.1 | - | 7 | 632 | |||
BC132475.1 | - | 7 | 599 | 40130752 | ||
BC137854.1 | - | 7 | 599 | 8860873 | ||
BC144975.1 | - | 7 | 599 | 9053523 |