Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145806.1 | NP_001139278.1 | 1 | 276 | ||
NM_130890.2 | NP_570960.2 | 1 | 276 | |||
GenBank mRNA | AB050202.1 | - | 1 | 272 | ||
AB050203.1 | - | 1 | 272 | |||
AB061518.1 | - | 1 | 222 | |||
AB061519.1 | - | 1 | 222 | |||
AK132449.1 | - | 1 | 277 | |||
BC125504.1 | - | 1 | 248 | 40130019 | ||
BC125508.1 | - | 1 | 248 | 40130023 | ||
BC145055.1 | - | 1 | 248 |