Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080729.3 | NP_542767.1 | 3 | 809 | ||
GenBank mRNA | AF458060.1 | - | 2 | 733 | ||
BC120568.1 | - | 3 | 808 | 40129491 | ||
BC125524.1 | - | 3 | 808 | 40130039 | ||
BC144962.1 | - | 3 | 808 | 9053510 |