Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080847.3 | NP_543123.2 | 2 | 223 | ||
GenBank mRNA | AF403043.1 | - | 2 | 224 | ||
AK019556.1 | - | 2 | 224 | |||
AK029759.1 | - | 2 | 155 | |||
AK079418.1 | - | 2 | 329 | |||
BC050794.1 | - | 2 | 172 | 6742480 |