Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080849.3 | NP_543125.1 | 2 | 290 | ||
GenBank mRNA | AF407579.1 | - | 2 | 240 | ||
AK014546.1 | - | 2 | 251 | |||
AK154358.1 | - | 2 | 253 | |||
BB646522.1 | - | 2 | 300 | |||
BC057995.1 | - | 2 | 235 | |||
BC070457.1 | - | 2 | 239 | 30603080 | ||
BY233061.1 | - | 2 | 292 |