Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267724.1 | NP_001254653.1 | 2 | 171 | ||
NM_178283.4 | NP_840068.1 | 4 | 510 | |||
GenBank mRNA | AF403041.1 | - | 4 | 510 | ||
AK078289.1 | - | 4 | 466 | |||
AK133892.1 | - | 4 | 436 | |||
BC018240.1 | - | 4 | 464 | 4239996 | ||
BG080214.2 | - | 2 | 105 |