Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267707.1 | NP_001254636.1 | 8 | 1012 | ||
NM_130861.3 | NP_570931.1 | 8 | 960 | |||
GenBank mRNA | AF240694.1 | - | 8 | 930 | ||
AK029706.1 | - | 8 | 961 | |||
AK166446.1 | - | 8 | 989 |