Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012450.1 | NP_001012453.1 | 17 | 2820 | ||
NM_001012451.1 | NP_001012454.1 | 16 | 2531 | |||
NM_080471.3 | NP_536719.2 | 16 | 2660 | |||
GenBank mRNA | AK133551.1 | - | 15 | 2547 | ||
AK173070.1 | - | 16 | 2571 | |||
AY026320.2 | - | 15 | 2453 | |||
BC065177.1 | - | 17 | 2770 | 6825961 |