Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080560.3 | NP_542127.1 | 4 | 1332 | ||
GenBank mRNA | AK005302.1 | - | 4 | 1350 | ||
AK150930.1 | - | 1 | 273 | |||
AK161968.1 | - | 4 | 1348 | |||
BC067069.1 | - | 4 | 1333 | 30550024 | ||
BC132630.1 | - | 4 | 1307 | 40130907 | ||
BC138211.1 | - | 4 | 1307 | 8861230 |