Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000578.3 | NP_000569.3 | 3 | 574 | ||
XM_005246793.3 | XP_005246850.1 | 2 | 694 | |||
XM_005246794.3 | XP_005246851.1 | 3 | 837 | |||
XM_006712709.3 | XP_006712772.1 | 2 | 694 | |||
XM_011511684.2 | XP_011509986.1 | 3 | 837 | |||
XM_011511685.2 | XP_011509987.1 | 2 | 694 | |||
XM_017004766.1 | XP_016860255.1 | 3 | 805 | |||
XM_017004767.1 | XP_016860256.1 | 3 | 401 | |||
GenBank mRNA | AB209650.1 | - | 2 | 1507 | ||
AK294707.1 | - | 3 | 315 | |||
AK298667.1 | - | 3 | 328 | |||
AK303398.1 | - | 3 | 298 | |||
AK307954.1 | - | 3 | 574 | |||
AK310914.1 | - | 3 | 330 | |||
AK311405.1 | - | 2 | 181 | |||
BC071165.1 | - | 3 | 294 | 5756382 | ||
D38171.1 | - | 1 | 81 | |||
D50402.1 | - | 3 | 409 | |||
D50403.1 | - | 3 | 320 | |||
L32185.1 | - | 3 | 310 |