Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267724.1 | NP_001254653.1 | 4 | 1897 | ||
NM_178283.4 | NP_840068.1 | 6 | 2372 | |||
GenBank mRNA | AK078289.1 | - | 6 | 2323 | ||
AK133892.1 | - | 6 | 2296 | |||
BC018240.1 | - | 6 | 2317 | 4239996 |