Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130452.1 | NP_569719.1 | 2 | 417 | ||
GenBank mRNA | AK050010.1 | - | 435 | |||
AY033514.1 | - | 2 | 355 | |||
BC019406.1 | - | 2 | 417 | 5134018 |