Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130858.3 | NP_570928.1 | 2 | 864 | ||
GenBank mRNA | AK038955.1 | - | 2 | 541 | ||
AK158760.1 | - | 2 | 865 | |||
BC007167.1 | - | 2 | 570 | 3490479 |