Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 4 | 424 | ||
GenBank mRNA | AF327431.1 | - | 4 | 296 | ||
AK089276.1 | - | 4 | 263 | |||
AK142397.1 | - | 2 | 459 | |||
AK143217.1 | - | 4 | 425 | |||
AK144607.1 | - | 4 | 405 | |||
AK155733.1 | - | 3 | 635 | |||
AK157340.1 | - | 4 | 396 | |||
BC003901.1 | - | 4 | 369 | 3601310 |