Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130163.1 | NP_001123635.1 | 8 | 1523 | ||
NM_001130165.1 | NP_001123637.1 | 8 | 1618 | |||
NM_001130166.1 | NP_001123638.1 | 10 | 1882 | |||
NM_130885.2 | NP_570955.1 | 8 | 1523 | |||
GenBank mRNA | AF324899.1 | - | 8 | 1522 | ||
AF333985.1 | - | 8 | 1523 | |||
AK040881.1 | - | 8 | 1546 | |||
AK158703.1 | - | 10 | 1864 | |||
BC003927.1 | - | 2 | 113 | |||
BC089183.1 | - | 8 | 1613 | 5321255 | ||
BC098491.1 | - | 10 | 1865 | 30086857 |