Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130893.3 | NP_570963.1 | 1, 2 | 547 | ||
GenBank mRNA | BC057562.1 | - | 1, 2 | 320 | 6401572 | |
BC089316.1 | - | 1, 2 | 331 | |||
CB525529.1 | - | 1, 2 | 547 |