Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290651.1 | NP_001277580.1 | 4 | 737 | ||
NM_001290652.1 | NP_001277581.1 | 4 | 737 | |||
GenBank mRNA | AF285577.1 | - | 6 | 563 | ||
AK016533.1 | - | 7 | 609 | |||
AK031490.1 | - | 4 | 716 | |||
AK040521.1 | - | 4 | 739 | |||
BC056382.1 | - | 4 | 427 | 6853936 |